Learn More
Thermo Scientific™ Remel™ Contact II Tryptic Soy Agar w/Lecithin, Polysorbate 80
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Remel Contact II Tryptic Soy Agar w/Lecithin, Polysorbate 80 is used for the cultivation of bacteria, yeast and fungi.
- Manufactured in a clean room environment to provide the highest level of sterility assurance
- Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
- Contact I (Single Bag) and Contact II (Double Bag) are manufactured in a clean room environment
- This provides the highest level of sterility assurance with no compromise of growth characteristics
- Each lot is tested to ensure consistent, reproducible results
Specifications
Specifications
| Description | Contact II Tryptic Soy Agar w/Lecithin, Polysorbate 80 |
| Format | Contact - Double Bag |
| Product Type | Agar Plate |
| Quantity | 100/Pk. |
Not intended for IVD use.