Learn More
Thermo Scientific™ Remel™ Contact Plate Sterile D/E (Dey-Engley) Neutralizing Agar (Irradiated)
Shop All Thermo Scientific Products
Description
Thermo Scientific™ Remel Contact Plate Sterile D/E Neutralizing Agar (Irradiated) is used for the cultivation of microorganisms with neutralization of specific chemical disinfectants.
- Contact Plates Sterile are double-bagged and a third bag is enclosed for customer use (Double + Bag)
- These products are manufactured in a clean room and validated sterile by gamma radiation
- The Contact Plates are made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Contact Plates, D/E-Neutralizing Agar |
| Format | Contact - Double Bag |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
Not intended for IVD use.