Learn More
Thermo Scientific™ Remel™ Contact Plate Sterile Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80 (pH 5.6) (Irradiated)
Catalog No. R111805 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
R111805 10/Pk.
1 options
Catalog No. R111805 Supplier Thermo Scientific™ Supplier No. R111805
Add to Cart
Add to Cart
Thermo Scientific™ Remel Contact Plate Sterile Sabouraud Dextrose Agar with Lecithin, Polysorbate 80 (pH 5.6, Irradiated) is used for cultivation of yeast and fungi.
Thermo Scientific™ Remel Contact Plate Sterile Sabouraud Dextrose Agar with Lecithin, Polysorbate 80 (pH 5.6) (Irradiated) is used for the cultivation of yeast and fungi.
  • Contact Plates Sterile are double bagged plus a 3rd bag is enclosed for customer use (Double + Bag)
  • These products are manufactured in a clean room and validated sterile by gamma radiation
  • The Contact Plates Sterile listed are made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
TRUSTED_SUSTAINABILITY
Specifications
Description Contact Plates; Sabouraud Dextrose Agar w/Lecithin; Gamma irradiated
Format Contact - Double Bag
Product Type Agar Plate
Quantity 10/Pk.
Not intended for IVD use.