Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel Cooked Meat Medium w/Glucose, Hemin, Vitamin K
GSA_VA

Catalog No. R060546 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 7 mL
100 x 7 mL
20 x 10 mL
Format:
15mm x 103mm Tube
16mm x 125mm Tube, Hungate Cap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R060546 20 x 7 mL 15mm x 103mm Tube
R060544 100 x 7 mL 15mm x 103mm Tube
R07086 20 x 10 mL 16mm x 125mm Tube, Hungate Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R060546 Supplier Thermo Scientific™ Supplier No. R060546
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Remel Cooked Meat Medium with Glucose, Hemin, Vitamin K is for the cultivation of anaerobic microorganisms.

Thermo Scientific™ Remel Cooked Meat Medium with Glucose, Hemin, Vitamin K is for the cultivation of anaerobic microorganisms.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Cooked Meat Medium w/Glucose, Hemin, Vitamin K
Format 15mm x 103mm Tube
Product Type Microbiology Media
Packaging Type 20/Pk.
Quantity 20 x 7 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.