Learn More
Thermo Scientific™ Cornmeal Agar with Polysorbate 80, Trypan Blue
Description
Cultivate fungi and stimulate chlamydospore production by Candida albicans with Cornmeal Agar with Polysorbate 80, Trypan Blue.
Cultivate fungi and stimulate chlamydospore production by Candida albicans with Thermo Scientific™ Remel™ Cornmeal Agar with Polysorbate 80, Trypan Blue. Polack and Benham demonstrated the use of cornmeal agar for studying differential mycelial characteristics of Candida species1,2. Adding polysorbate 80 to cornmeal agar stimulates chlamydospore formation by Candida albicans3-4.
Cornmeal Agar with Polysorbate 80 and trypan blue is a solid medium recommended for use in qualitative procedures as a general-purpose media for cultivating fungi and stimulating chlamydospore production by Candida albicans.
- Ready-to-use: Convenience of prepared media.
- Easy-to-read: Trypan blue provides a contrasting background for the optimal visualization of chlamydospores.
- General-purpose media: Helps cultivate fungi and stimulate chlamydospore formation by Candida albicans.
Cornmeal agar consists of an infusion of cornmeal and agar, which provides the essential nutrients needed to support the growth of fungi and the production of chlamydospores by Candida albicans. Polysorbate 80 reduces surface tension and enhances the development of chlamydospores. Trypan blue provides a contrasting background allowing optimal visualization of chlamydospores.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | Cornmeal Agar w/Tween™ 80, Trypan Blue |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.