Learn More
Thermo Scientific™ Cornmeal Agar with Polysorbate 80 with or without Trypan Blue
Shop All Thermo Scientific Products
Description
Cultivate fungi and stimulate chlamydospore production by Candida albicans with this ready-to-use medium.
Cultivate fungi and stimulate chlamydospore production by Candida albicans with Thermo Scientific™ Remel Cornmeal Agar with Polysorbate 80 general purpose medium, with or without Trypan Blue. Polack and Benham demonstrated the use of cornmeal agar for studying differential mycelial characteristics of Candida species1,2. Adding polysorbate 80 to cornmeal agar stimulates chlamydospore formation by Candida albicans3-4.
Cornmeal Agar with Polysorbate 80 with or without trypan blue is a solid medium recommended for use in qualitative procedures as a general-purpose media for cultivating fungi and stimulating chlamydospore production by Candida albicans.
- General-purpose media: cultivate fungi and stimulate chlamydospore formation by Candida albicans.
- Easy-to-read: Trypan blue provides a contrasting background for the optimal visualization of chlamydospores.
- Ready-to-use: the convenience of prepared media.
Cornmeal agar consists of an infusion of cornmeal and agar, which provides the essential nutrients needed to support the growth of fungi and the production of chlamydospores by Candida albicans. The agar is a solidifying agent. Polysorbate 80 reduces surface tension and enhances the development of chlamydospores. Trypan blue is an optimal additive that provides a contrasting background, thereby permitting optimal visualization of chlamydospores.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Cornmeal Agar w/Tween™ 80 |
| Format | Monoplate Deep Pour |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |