Learn More
Description
Detect protozoan cysts and helminth eggs and larvae in wet mount preparations using D'Antoni's Iodine Stain.
Detect protozoan cysts and helminth eggs and larvae in wet mount preparations using Thermo Scientific™ Remel™ D'Antoni's Iodine stain. This is a nonspecific, contrast dye, added to fresh, unpreserved fecal specimens to aid in detection of helminth eggs and larvae and in differentiating protozoan cysts from host white blood cells1,2.
The diagnosis of intestinal parasitic infections is confirmed by the identification of parasites in fecal specimens. D’Antoni’s Iodine stains the nuclei of protozoan cysts, making them more visible. Helminth eggs and larvae are recognizable in wet mounts, though iodine staining occasionally obscures the internal morphologic details.
- Ready-to-use: Liquid reagent supplied in 25mL dropper bottles
- Easy identification: Stains the nuclei of protozoan cysts, making them more visible.
- Easy-to-store: Store at room temperature (20-25°C)
D’Antoni’s Iodine preparation contains 15g iodine and 10g potassium iodide dissolved in demineralized water. This preparation is ready to use and does not require further dilution.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Content And Storage | Store at Room Temperature (20-25 °C) Until Used |
| Description | D'Antoni's Iodine |
| Format | Bottle |
| Packaging Type | 25 mL/Bottle, Ea. |
| Quantity | 25 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.