Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Dermatophyte Test Medium (DTM)
GSA_VA

Catalog No. R01365 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
20/Pk.
Format:
F - slant
Monoplate Deep Pour
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R01365 10/Pk. Monoplate Deep Pour
R08352 20/Pk. F - slant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R01365 Supplier Thermo Scientific™ Supplier No. R01365
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Facilitate selective and differential isolation of pathogenic fungi (dermatophytes) from cutaneous sources using Dermatophyte Test Medium (DTM).

Facilitate selective and differential isolation of pathogenic fungi (dermatophytes) from cutaneous sources using Thermo Scientific™ Remel™ Dermatophyte Test Medium (DTM). This medium was formulated by Taplin et al. for use in locations where specialized training and microscopic examination is not available1. A pH indicator and three antimicrobial agents are added to the agar to provide a differential and selective medium for isolation of dermatophytes belonging to the genera Microsporum, Trichophyton, or Epidermophyton.

Contains a pH indicator.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Dermatophyte Test Medium
Format Monoplate Deep Pour
Product Type Agar
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.