Learn More
Thermo Scientific™ Dermatophyte Test Medium (DTM)
Description
Facilitate selective and differential isolation of pathogenic fungi (dermatophytes) from cutaneous sources using Dermatophyte Test Medium (DTM).
Facilitate selective and differential isolation of pathogenic fungi (dermatophytes) from cutaneous sources using Thermo Scientific™ Remel™ Dermatophyte Test Medium (DTM). This medium was formulated by Taplin et al. for use in locations where specialized training and microscopic examination is not available1. A pH indicator and three antimicrobial agents are added to the agar to provide a differential and selective medium for isolation of dermatophytes belonging to the genera Microsporum, Trichophyton, or Epidermophyton.
Specifications
Specifications
| Description | Dermatophyte Test Medium |
| Format | F - slant |
| Product Type | Culture Medium |
| Quantity | 20/Pk. |