Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Dermatophyte Test Medium (DermaTube)
GSA_VA

Catalog No. R241045 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 12 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R241045 10 x 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R241045 Supplier Thermo Scientific™ Supplier No. R241045

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Facilitate selective and differential isolation of pathogenic fungi from cutaneous sources using Dermatophyte Test Medium (DermaTube).

Facilitate selective and differential isolation of pathogenic fungi from cutaneous sources using Thermo Scientific™ Remel™ Dermatophyte Test Medium (DermaTube). This medium was formulated by Taplin et al. for use in locations where specialized training and microscopic examination is not available1. A pH indicator and three antimicrobial agents are incorporated into the agar to provide a differential and selective medium for isolation of dermatophytes belonging to the genera Microsporum, Trichophyton, or Epidermophyton.

The dermatophytes are fungi that possess keratinolytic properties that enable them to invade skin, nails and hair2. The infections caused by these organisms are commonly referred to as ringworm and are classified by the Latin word tinea followed by the area of the body infected3. Facilitate selective and differential isolation of pathogenic fungi from cutaneous sources using Dermatophyte Test Medium (DermaTube).

  • Ready to Use: Convenience of prepared media.
  • Selective: Cyclohexamide, chloramphenicol, and gentamycin are selective agents which inhibit most saprophytic fungi and many gram-positive and gram-negative bacteria, including some Pseudomonas.
  • Ease to identify: Phenol red is a pH indicator which detects alkaline metabolites produced by dermatophytes, resulting in a red color development of the medium.

This medium is recommended for use in qualitative procedures. This medium contains soy peptones that supply the nitrogen and carbon compounds necessary for the growth of microorganisms. Dextrose is an energy source.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description DermaTube™
Format 27mm x 70mm Tube
Product Type Microbiology Media
Quantity 10 x 12 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.