Learn More
Thermo Scientific™ Diamond’s Medium Modified
Catalog No. R07097 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
Format:
E - 10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R07097 20/Pk. E - 10mL
R07096 100/Pk. E - 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R07097 Supplier Thermo Scientific™ Supplier No. R07097
Add to Cart
Add to Cart
This item is not returnable. View return policy
Facilitates the isolation of Trichomonas vaginalis from clinical specimens using Diamond's Medium Modified.
Facilitates the isolation of Trichomonas vaginalis from clinical specimens using Thermo Scientific™ Remel™ Diamond's Medium Modified. Diamond's medium was developed by Diamond in 1957 for clinical diagnostic use1. In 1980, The Centers for Disease Control and Prevention modified the original Diamond's Medium by replacing sheep serum with horse serum, increasing the concentration of maltose, cysteine, and ascorbic acid, eliminating agar, and adding antibiotics to suppress the growth of bacteria and fungi2.
Facilitate the isolation of Trichomonas vaginalis from clinical specimens using Diamond’s Medium Modified.
  • Ready to Use: Convenience of prepared media.
  • Less Sample Required: T. vaginalis can be recovered from an inoculum containing as few as a single organism.
  • Selective: Penicillin, streptomycin and amphotericin B collectively supress the growth of a wide range of urogenital flora without inhibiting the growth of T. vaginalis.
This medium is recommended for use in qualitative procedures. This medium contains caesin peptones, cystein, and yeast extract that provide amino acid, nitrogen, sulfur, carbon and vitamins to support the growth of T. vaginalis. Maltose is an energy source and horse serum provides essential growth factors.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Diamond's Medium Modified
Format E - 10mL
Product Type Microbiology Media
Quantity 20/Pk.