Learn More
Thermo Scientific™ Remel™ DMSO Modified Acid Fast Stain Kit
Catalog No. 23313392 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
3/Kit
Catalog No. Quantity
23-313-392 3/Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-313-392 Supplier Thermo Scientific™ Supplier No. R246303
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel DMSO Modified Acid Fast Stain Kit is for detection of acid-fast microorganisms.
Thermo Scientific™ Remel DMSO Modified Acid Fast Stain Kit is for detection of acid-fast microorganisms.
Includes: 1 bottle Reagent A Fuchsin DMSO Stain; 1 bottle Reagent B Decolorizer; 1 bottle Reagent C Counterstain.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 3/Kit
Includes 1 bottle Reagent A Fuchsin DMSO Stain; 1 bottle Reagent B Decolorizer; 1 bottle Reagent C Counterstain.
Volume (Metric) 250 mL/Bottle
Product Type Acid Fast Stain