Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EC Medium w/MUG

Catalog No. R07657 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
16mm x 125mm Durham Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R07657 16mm x 125mm Durham Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R07657 Supplier Thermo Scientific™ Supplier No. R07657

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect Escherechia coli in water and food samples by a fluorogenic assay procedure with EC Medium w/MUG.

Detect coliforms by fluorogenic assay procedure from water, wastewater, and food samples with Thermo Scientific™ Remel™ EC Medium w/MUG.

Hajna and Perry developed EC Medium to detect Escherichia coli and other coliforms1 in water, milk, shellfish, and other materials2. Feng and Hartman added 4-methylumbelliferyl-β-D-glucuronide (MUG)3 to conventional coliform assays. This improved detection rates of E. coli as fluorescence could be detected within 4-24 hours.

EC Medium w/MUG is a liquid medium used to isolate and detect Escherechia coli. E. coli is an indicator organism that can indicate fecal contamination or unsanitary conditions.

  • Ready-to-use: Convenience of prepared media.
  • Selective: Bile inhibits some gram-positive cocci and sporeformers.
  • Easy-to-detect: Improved detection rates of E. coli due to fluorescence of 4-methylumbelliferone within 4-24 hours.
  • Recommended by American Public Health Association (APHA) and Food and Drug Administration (FDA)4,5 for detection of coliforms in water, wastewater and food.

EC Medium w/MUG contains casein and meat peptones that supply amino acids, peptides and nitrogenous compounds required for growth of bacteria. Lactose acts as a ready source of energy and its fermentation leads to production of gas in the vial. Sodium chloride provides essential electrolytes and maintains osmotic equilibrium. Dipotassium phosphate and monopotassium phosphate are buffers which help control the pH. MUG is hydrolyzed by glucuronidase and yields a fluorescent end product, 4-methylumbelliferone.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM.
Description EC Medium w/MUG
Format 16mm x 125mm Durham Tube
Product Type Prepared Microbiology Tubes
Quantity 20 x 10 mL

Not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.