Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Egg Yolk Tellurite 100%

Catalog No. R450330
Change view
Click to view available options
Quantity:
100 mL
Packaging Type:
1 Ea.
10/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
R450330 100 mL 1 Ea.
R450331 100 mL 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R450330 Supplier Thermo Scientific™ Supplier No. R450330
Add to Cart
Add to Cart
This item is not returnable. View return policy

Remel Egg Yolk Tellurite 100% is used with Baird Parker Agar Base.

Remel Egg Yolk Tellurite 100% is used with Baird Parker Agar Base.

TRUSTED_SUSTAINABILITY

Specifications

Description Egg Yolk Tellurite 100%
Form Liquid
Product Type Microbiology Media
Packaging Type 1 Ea.
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.