Learn More
Thermo Scientific™ Remel™ Enhanced Gram Iodine
GSA_VA
Catalog No. R40234 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5/Pk.
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R40234 5/Pk.
R40235 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R40234 Supplier Thermo Scientific™ Supplier No. R40234
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ enhanced gram stain reagents are balanced to give a distinct gram-positive reaction and are less prone to over decolorization.
Thermo Scientific™ Remel enhanced gram stain reagents are balanced to give a distinct gram-positive reaction and are less prone to over decolorization.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5/Pk.
Certifications/Compliance CE Marked
Volume (Metric) 5 x 250 mL
Product Type Gram Iodine