Learn More
Thermo Scientific™ Remel™ Enhanced Gram Stain Kit
GSA_VA
Catalog No. R40240 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Kit
Catalog No. Quantity
R40240 Kit
1 options
This item is not returnable. View return policy
Catalog No. R40240 Supplier Thermo Scientific™ Supplier No. R40240
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ enhanced gram stain reagents are balanced to give a distinct gram-positive reaction and are less prone to over decolorization.
Thermo Scientific™ Remel enhanced gram stain reagents are balanced to give a distinct gram-positive reaction and are less prone to over decolorization.
  • Gram Enhancer is used to reduce the red color of background debris to give better contrast to small gram-negative microorganisms
Includes:
1 Gram Crystal Violet, Enhanced; 1 Gram Iodine, Enhanced; 1 Gram Decolorizer/Fixative, Enhanced; 1 Gram Safranin, Enhanced; 1 Gram Enhancer.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Quantity Kit
Includes 1 Gram Crystal Violet, Enhanced; 1 Gram Iodine, Enhanced; 1 Gram Decolorizer/Fixative, Enhanced; 1 Gram Safranin, Enhanced; 1 Gram Enhancer.
Volume (Metric) 250 mL/Bottle
Product Type Gram Stain Kit