Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Esculin Hydrolysis Agar
GSA_VA

Catalog No. R060878 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20/Pk.
Format:
15mm x 103mm Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R060878 20/Pk. 15mm x 103mm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R060878 Supplier Thermo Scientific™ Supplier No. R060878
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect esculin hydrolysis by aerobic and anaerobic microorganisms with Esculin Hydrolysis Agar.

Differentiate gram positive and gram negative bacteria on the basis of their ability to hydrolyze esculin with Thermo Scientific™ Remel™ Esculin Hydrolysis Agar.

Harisson and Van der Leck observed the ability of organisms to hydrolyze esculin in the year 19091. In 1924, Rochaix used esculin hydrolysis to identify group D streptococci2. Smith later suggested that esculin hydrolysis test can be used to differentiate certain anaerobic bacteria3. In 1974, Dowell et al. used the esculin hydrolysis test to differentiate Bacteroides species4.

Esculin Hydrolysis Agar is a solid medium used to detect esculin hydrolysis by microorganisms. The esculin hydrolysis test is used for identifying a wide variety of microorganisms, including Enterobacteriaceae, Streptococcus and Listeria, non-fermentative gram-negative bacilli, and anaerobes.

Esculin is enzymatically hydrolyzed by bacteria and yeilds esculetin and glucose. Esculetin combines with ferric ions to produce a brownish-black colored compound5. Esculin hydrolysis can thus be determined by detecting the end product esculetin using Esculin Hydrolysis Agar.

  • Ready-to-use: convenience of prepared media.
  • Easy-to-interpret:
    • Positive test- brownish-black color development.
    • Negative test- no color change.

Esculin Hydrolysis Agar contains beef heart infusion, casein peptone and yeast extract that supply amino acids, vitamins, and trace minerals necessary for growth. Sodium chloride provides essential electrolytes and maintains osmotic equilibrium. Vitamin K and hemin are growth factors that enhance growth of anaerobic bacteria. Agar is a solidifying agent.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Esculin Hydrolysis Agar
Format 15mm x 103mm Tube
Product Type Agar Plate
Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.