Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Fastidious Broth

Catalog No. R07664 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R07664 100 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R07664 Supplier Thermo Scientific™ Supplier No. R07664
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivate fastidious organisms present in small numbers from clinical specimens with Fastidious Broth.

Cultivate fastidious organisms present in small numbers from clinical specimens with Thermo Scientific™ Remel™ Fastidious Broth.

In 1994 Cartwright et al. first described the use of Fastidious Broth to enrich specimens that may contain small numbers of fastidious organisms, prior to culture on plated media1. Fastidious Broth support the growth of a variety of bacteria e.g., Corynebacterium jeikeium, Haemophilus influenzae, Neisseria gonorrhoea and Streptococcus pneumoniae. For many organisms this broth reduces the time of incubation and the volume of inoculum required for growth to be detectable.

Fastidious Broth is a liquid medium recommended for cultivation of fastidious organism. Fastidious Broth contains casein and meat peptones that supply amino acids, peptides and nitrogenous substances essential for growth. Yeast extract supplies essential B-complex vitamins that stimulate growth of organisms. Sodium chloride provides essential electrolytes which maintain osmotic equilibrium.

  • Ready-to-use: Convenience of prepared media.
  • Easy-to-use: Inoculate the broth directly with specimen immediately once it is received in the laboratory and incubate at 35-37º C for up to 7 days.

Dextrose is a carbohydrate source that provides energy. L-cysteine improves recovery of aerobic and anaerobic microorganisms from blood. Tris is a buffer. Magnesium and iron salts improve the growth of aerobes and anaerobes. NAD and hemin help in growth of certain fastidious organism that include H. influenzae. Polysorbate 80 is the nutritional source of essential fatty acids. Agarose is added to hinder diffusion of oxygen. Pyridoxal is essential for the growth of many fastidious organisms.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Fastidious Broth
Format 16mm x 125mm Tube
Product Type Microbiology Media
Quantity 100 x 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.