Learn More
Thermo Scientific™ Fecal Concentrator Kit II
Catalog No. R21911 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 Tests/Kit
Catalog No. Quantity
R21911 50 Tests/Kit
1 options
This item is not returnable. View return policy
Catalog No. R21911 Supplier Thermo Scientific™ Supplier No. R21911
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Concentrates the protozoan cysts, oocysts, and helminth eggs and larvae from fecal material using Fecal Concentrator Kit II.
Concentrate the protozoan cysts, oocysts, and helminth eggs and larvae from fecal material using Thermo Scientific™ Remel™ Fecal Concentrator Kit II. This kit utilizes existing sedimentation methodology along with standardized components to achieve recovery of protozoan cysts, oocysts and helminth eggs and larvae from preserved fecal specimens. The filtration assembly includes a filter cap screwed to a graduated 50 mL centrifuge tube. Pore diameters in the filters are 600 microns, allowing passage of cysts, eggs and larvae while retaining macroscopic fecal matter and debris on top.1
The diagnosis of intestinal parasitic infections requires appropriate collection, transportation, concentration, staining and identification of parasites from fecal specimens4,5.
This kit filters the concentrate of protozoan cysts, oocysts, and helminth eggs and larvae from fecal material.
  • Compatible with most fecal transport vials
  • Less sample required
This kit utilizes existing sedimentation methodology along with standardized components to achieve recovery of protozoan cysts, oocysts and helminth eggs and larvae from preserved fecal specimens. The filtration assembly includes a filter cap screwed to a graduated 50 mL centrifuge tube. Pore diameters in the filters are 600 microns, allowing passage of cysts, eggs and larvae while retaining macroscopic fecal matter and debris on top6. The surfactant, 10% Triton X-100, reduces surface tension to enhance filtration and frees additional helminth eggs that may be trapped in fecal debris or mucus. Ethyl acetate or Xylene substitutes dissolve any fat present in the specimen and aid in the separation of fecal debris from concentrated sediment containing parasites. Ideally 0.5 to 1.0 mL of fecal sediments should remain for adequate recovery and examination of parasites.
The most current sedimentation techniques involve filtering, centrifugation, and defatting steps. The earlier used Ritchie formalin-ether sedimentation technique has been modified, in which ether is replaced with other defatting agents such as ethyl acetate or xylene substitutes to avoid safety issues associated with ether2-4.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fecal Concentrator Kit
Description Fecal Concentrator Kit II
Quantity 50 Tests/Kit