Learn More
Thermo Scientific™ Remel™ Fecal Concentrator Kit
GSA_VA
Catalog No. R21901 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
36 Tests/Kit
Catalog No. Quantity
R21901 36 Tests/Kit
1 options
This item is not returnable. View return policy
Catalog No. R21901 Supplier Thermo Scientific™ Supplier No. R21901
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel Fecal Concentrator Kit is used for concentration of intestinal parasites from preserved fecal specimens.
Thermo Scientific™ Remel Fecal Concentrator Kit is used for concentration of intestinal parasites from preserved fecal specimens.
Includes:
36 Filtration funnels, 36 graduated conical tubes and caps, 1 vial Triton™ X-100 mucolytic reagent
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fecal Concentrator Kit
Description Fecal Concentrator Kit
Quantity 36 Tests/Kit