Learn More
Thermo Scientific™ Fecal Transport Three-Vial Sets: Cary-Blair w/Indicator/SAF/Empty Vial
Shop All Thermo Scientific ProductsDescription
Transport, preserve, and examine stool specimens for enteric pathogens using Remel™ Fecal Transport Three-Vial Sets.
Transport, preserve, and examine stool specimens for enteric pathogens using Thermo Scientific™ Remel™ Fecal Transport Three-Vial Sets containing Cary-Blair with Indicator/SAF Fixative/Empty Vials. The Cary-Blair formulation has a low nutrient content, a low oxidation-reduction potential, and a high pH and recovers Shigella and Salmonella1. Luechtefeld et al. modified this medium and reduced agar content2,3 to recover Campylobacter spp. It is also recommended for the transport of Yersinia species and Vibrio parahaemolyticus4,5.
Cary Blair Transport Medium with Indicator is an isotonic and non-nutritive medium and does not contain a fermentable carbohydrate. SAF (Sodium acetate-acetic acid-formalin) Fixative is a multipurpose medium used for preservation of intestinal parasites6. Remel™ Fecal Transport Three-Vial Sets are packaged in zip seal bags with multilingual instructions.
- Caps are color coded to match industry standard colors and have an attached spork.
- Broad range with long shelf life
- Industry standard color-coded caps and labels
- Easy to use spork cap for safe collection
- Convenient packaging, including bulk kits
- Multilingual patient collection instructions
- Empty vial is used for test procedures not compatible with fecal preservatives
- Cary-Blair with indicator contains 0.16% agar and is for preservation and recovery of enteric bacterial pathogens
- SAF fixative provides preservation, concentration and permanent staining of intestinal parasites
Cary-Blair with indicator medium contains sodium thioglycolate which is added to impede oxidation. Disodium phosphate is incorporated in the medium to act as a buffering agent, that prevents overgrowth of Escherichia coli and Enterobacter species. The relatively high pH due to acid formation minimizes the destruction of bacteria. Phenol red is a pH indicator which is added to demonstrate acidic shifts that can affect the viability of some enteric pathogens.
SAF Fixative can be used in multiple procedures such as wet mount examination, concentration method, and permanent stained smears7,8. SAF is a safer than Schaudinn’s fixative9 as it does not contain mercuric chloride. Microscope slides used to prepare permanent stains should be coated with albumin before application of stool sample that has been preserved in SAF Fixative10.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Product Type | Fecal Transport Set |
| Description | PVA, Low Viscosity/Formalin 10%/Cary Blair; SAF/Empty Vial; 3-vial Sets; 84 Sets/Pk. |
| Packaging | 3-Vial Set |
| Quantity | 84 Sets/Pk. |
| Volume (English) | 1.01 oz. |
| Volume (Metric) | 30 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.