Learn More
Thermo Scientific™ Remel™ Fecal Transport Two-Vial Sets: Cary-Blair w/Indicator/Empty Vial
Catalog No. R21616 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
15 mL
30 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
R21616 15 mL
R21949 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R21616 Supplier Thermo Scientific™ Supplier No. R21616
Add to Cart
Add to Cart
This item is not returnable. View return policy
Remel Fecal Transport Two-Vial Sets: Cary-Blair w/Indicator/Empty Vial offer preservation and recovery of enteric bacterial pathogens.
Remel Fecal Transport Two-Vial Sets: Cary-Blair w/Indicator/Empty Vial offer preservation and recovery of enteric bacterial pathogens.
Caps are color coded to match industry standard colors and have an attached spork.
  • Broad range with long shelf life
  • Industry standard color-coded caps and labels
  • Easy to use spork cap for safe collection
  • Convenient packaging, including bulk kits
  • Multilingual patient collection instructions
  • Empty Vial is used for test procedures not compatible with fecal preservatives
  • Cary-Blair with Indicator contains 0.16% agar and is for preservation and recovery of enteric bacterial pathogens
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fecal Transport Set
Description Cary-Blair/Empty Vial; Two-vial Sets
Packaging 2-Vial Set
Volume (Metric) 15 mL