Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Fecal Transport Two-Vial Sets: Cary-Blair w/Indicator/Empty Vial

Catalog No. R21949
Change view
Click to view available options
Volume (Metric):
15 mL
30 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
R21949 30 mL
R21616 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R21949 Supplier Thermo Scientific™ Supplier No. R21949
Add to Cart
Add to Cart
This item is not returnable. View return policy

Remel Fecal Transport Two-Vial Sets: Cary-Blair w/Indicator/Empty Vial offer preservation and recovery of enteric bacterial pathogens.

Remel Fecal Transport Two-Vial Sets: Cary-Blair w/Indicator/Empty Vial offer preservation and recovery of enteric bacterial pathogens.

Caps are color coded to match industry standard colors and have an attached spork.
  • Broad range with long shelf life
  • Industry standard color-coded caps and labels
  • Easy to use spork cap for safe collection
  • Convenient packaging, including bulk kits
  • Multilingual patient collection instructions
  • Empty Vial is used for test procedures not compatible with fecal preservatives
  • Cary-Blair with Indicator contains 0.16% agar and is for preservation and recovery of enteric bacterial pathogens
TRUSTED_SUSTAINABILITY

Specifications

Product Type Fecal Transport Set
Description Cary-Blair/Empty Vial; Two-vial Sets
Packaging 2-Vial Set
Volume (English) 1.01 oz.
Volume (Metric) 30 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.