Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Fluid K

Catalog No. R112332 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R112332 10 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R112332 Supplier Thermo Scientific™ Supplier No. R112332
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use as diluting or rinsing fluid for sterility testing on samples containing petrolatum.

Thermo Scientific™ Remel Fluid K (with 0.5% peptone, 0.3% beef extract, 1% Polysorbate 80 in water) is used as diluting or rinsing fluid for sterility testing on samples containing petrolatum.

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Buffer Solution
Packaging Type SB - 100mL
Quantity 10 x 100 mL

Not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.