Learn More
Thermo Scientific™ GCHI Enrichment with Rehydrating Solution
GSA_VA
Catalog No. R450411 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
Catalog No. Quantity
R450411 10 mL
1 options
This item is not returnable. View return policy
Catalog No. R450411 Supplier Thermo Scientific™ Supplier No. R450411
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cultivate fastidious microorganisms with GCHI Enrichment with Rehydrating Solution when used with GC Agar Base (R453502).
Cultivate fastidious microorganisms with Thermo Scientific™ Remel™ GCHI Enrichment with Rehydrating Solution when used with GC Agar Base (R453502).
Early culture methods for Neisseria gonorrhoeae included chocolate agar and the use of commercial peptones and yeast extract.
Various investigators studied nutritional requirements of gonococcus and found that components such as yeast extract, glutamine, cocarboxylase and cystine enhance the development of gonococci. The GCI Enrichment is used to cultivate pathogenic Neisseria. It has broad applications for other organisms as well, such as Haemophilus.
Cultivate fastidious microorganisms using GCHI Enrichment (lyophilized) with Rehydrating Solution.
  • Improve recovery time: GC Agar Base enriched with haemoglobin and yeast concentrate improves development of gonococci.
GCHI Enrichment with Rehydrating Solution contains vitamins, amino acids, coenzymes, dextrose, ferric ions, and other factors that enhances the growth of pathogenic Neisseria. Nicotinamide adenine dinucleotide (NAD) supplies enrichment necessary for the growth of Haemophilus spp.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description GCHI Enrichment w/Rehydrating Solution
Form Liquid
Product Type Enrichment
Quantity 10 mL