Learn More
Thermo Scientific™ Remel™ Giemsa Plus Stain Kit
Catalog No. 23313397 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
3/Pk.
Catalog No. Quantity
23-313-397 3/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-313-397 Supplier Thermo Scientific™ Supplier No. R246403
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel Giemsa Plus Stain Kit is a modified Wright stain for detection and identification of blood and tissue parasites.
Thermo Scientific™ Remel Giemsa Plus Stain Kit is a modified Wright stain for detection and identification of blood and tissue parasites.
Includes: 1 bottle Giemsa Plus Fixative; 1 bottle Giemsa Plus Reagent A; 1 bottle Giemsa Plus Reagent B.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 3/Pk.
Volume (Metric) 250 mL/Bottle
Product Type Giemsa Plus Stain Kit