Learn More
Thermo Scientific™ Remel™ GN Broth (Hajna) Media for Automation (MFA) Tubes
Shop All Thermo Scientific ProductsDescription
Take full advantage of your automated specimen processing systems with Thermo Scientific™ Remel™ GN Broth (Hajna) Media for Automation (MFA) Tubes. Formulated with the same high quality media used in traditional glass tubes, these plastic tubes are specifically designed to work with automated specimen processors. Maximize laboratory productivity and reduce the risk of human error when selectively enriching and isolating Gram-negative bacilli, especially Salmonella spp. and Shigella spp., by introducing complete specimen automation.
Efficiently isolate enteric Gram-negative bacilli, specifically Salmonella spp. and Shigella spp., from clinical specimens with Remel GN Broth (Hajna) Media for Automation (MFA) Tubes. GN Broth inhibits gram-positive organisms and enriches gram-negative organisms.
- Convenient: skirted design enables automated loading into specimen processors
- Consistent: the traditional broth formula you are familiar with
- Complementary: part of an extensive portfolio for your isolation, identification and differentiation workflow
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Cap Color | Yellow |
| Description | GN Broth (Hajna) Media for Automation (MFA) Tubes |
| Format | 15 x 103mm Tube |
| Product Type | Prepared Microbiology Tubes |
| Fill Volume | 4 mL |
| Quantity | 100/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.