Learn More
Description
Thermo Scientific™ Remel Graduated Conical Tube is for use with Remel Fecal Concentrator Filtration Device, Mfr. No. R21900.
- For use with Remel Fecal Concentrator Filtration Device
Specifications
Specifications
| Product Type | Conical Tube |
| Format | Tube |
| Description | Graduated Conical Tube |
| For Use With (Application) | Remel Fecal Concentrator Filtration Device Mfr. No. R21900 |
| Quantity | 1000/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.