Learn More
Thermo Scientific™ Remel™ Gram Crystal Violet
Catalog No. R40053 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 gal.
250 mL
250 mL, 5/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R40053 250 mL, 5/Pk.
R40052 250 mL
R40073 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R40053 Supplier Thermo Scientific™ Supplier No. R40053
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel Gram Crystal Violet is a primary stain in Gram stain procedures.
Thermo Scientific™ Remel Gram Crystal Violet is a primary stain in Gram stain procedures.
  • For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
  • Selected Gram stains are CE marked
TRUSTED_SUSTAINABILITY
Specifications
Quantity 250 mL, 5/Pk.
Certifications/Compliance CE Marked
Volume (Metric) 5 x 250 mL
Product Type Crystal Violet