Learn More
Thermo Scientific™ Remel™ Gram Iodine PVP
Description
Thermo Scientific™ Remel Gram Iodine PVP differs from traditional Gram stain formulation by substituting PVP iodine, an organic iodine complex, which is more stable than iodine.
- For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
- Selected Gram stains are CE marked
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | Each |
| Certifications/Compliance | CE Marked |
| Volume (Metric) | 3.785 L |
| Product Type | Gram Iodine |