Learn More
Thermo Scientific™ Haemophilus Isolation Agar w/bacitracin and horse blood
Shop All Thermo Scientific Products

Description
Selectively isolate Haemophilus species from respiratory specimens with Thermo Scientific™ Remel™ Haemophilus Isolation Agar w/bacitracin and horse blood. Haemophilus influenzae, a fastidious gram-negative bacillus, is a common cause of serious diseases of the upper and lower respiratory tract1.
Haemophilus Isolation Agar w/bacitracin and horse blood is a solid media recommended for use in qualitative procedures for selective isolation of Haemophilus spp. from respiratory specimens.
- Selective: Kanamycin and vancomycin antibiotics inhibit gram-positive organisms and facultative anaerobic bacteria. Neomycin stimulates the growth of gram-negative anaerobes, especially Bacteriodes spp. Colistin and nalidixic inhibit the growth of most gram-negative bacilli.
- Enriched medium: The presence of intact erythrocytes in horse blood enables differentiation of hemolytic and nonhemolytic Haemophilus spp. Horse blood, unlike sheep blood, does not contain V factor-inactivating enzymes.
- Ready-to-use: Convenience of prepared media.
Beef heart Infusion and casein peptone supply the nutritional requirements for all but the fastidious bacteria. Horse sheep supplies both the X and V factors, which are growth requirements for certain organisms, such as Haemophilus influenzae. Horse blood also serves to distinguish the hemolytic species of Haemophilus from non-hemolytic Haemophilus species. Bacitracin inhibits normal flora found in respiratory cultures, particularly Neisseria spp.4.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Haemophilus Isolation Agar w/bacitracin, horse blood |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.