Learn More
Thermo Scientific™ Haemophilus Test Medium (HTM)
Catalog No. R04033 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
15mm x 100mm Plate
15mm x 150mm Plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R04033 15mm x 150mm Plate
R01503 15mm x 100mm Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R04033 Supplier Thermo Scientific™ Supplier No. R04033
Add to Cart
Add to Cart
This item is not returnable. View return policy
Perform effortless susceptibility testing of Haemophilus influenza with this ready-to-use medium which conforms to CLSI standards.
Perform effortless susceptibility testing of Haemophilus influenza with ready-to-use Thermo Scientific™ Remel™ Haemophilus Test Medium (HTM). The medium forms part of the recommended methods by the Clinical and Laboratory Standards Institute (CLSI)1. Haemophilus Test Medium has been specifically formulated for the susceptibility testing of Haemophilus influenzae. The results achieved using HTM have been found to be highly reproducible2,3. The transparency of the medium allows zones of inhibition to be easily read through the bottom of the plate.
The occurrence of resistant strains of Haemophilus influenza has increased in recent decades, underscoring the need for reliable susceptibility testing methods for this organisms. Modifications to standard susceptibility testing procedures have become necessary because of the fastidious nature of H. influenza. Complex growth-stimulating supplements, ordinarily used to grow Haemophilus spp., have been found to promote antagonisms of certain antibiotics and result in poor reproducibility4. Difficulties in interpreting inhibition zones may also arise.
Use Haemophilus Test Medium (HTM) for the susceptibility testing of Haemophilus influenzae. 
  • Conforms to CLSI standards
  • Produces highly reproducible results.
  • Transparent medium to facilitate the reading of zones of inhibition.
  • Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Haemophilus Test Medium
Format 15mm x 150mm Plate
Product Type Agar Plate
Quantity 10/Pk.