Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HI Broth w/6.5% NaCl, BCP (5mL)
GSA_VA

Catalog No. R061046 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R061046 20 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R061046 Supplier Thermo Scientific™ Supplier No. R061046
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differentiate Enterobacteriaceae from group D Streptococci using Heart Infusion Broth with 6.5% NaCl (with Brom Cresol Purple) (Salt Broth).

Differentiate Enterobacteriaceae from group D streptococci using Thermo Scientific™ Remel™ Heart Infusion Broth with 6.5% NaCl (with Brom Cresol Purple) (Salt Broth). Facklam demonstrated that the differentiation of enterococci and nonenterococcal group D streptococci was more accurate on using Heart Infusion Broth with 6.5% NaCl Modified with Indicator in combination with bile and esculin agar1,2.

This media differentiates Enterobacteriaceae from group D streptococci. Enterococci grow in the presence of 6.5% NaCl and utilize dextrose in the medium. This medium contains beef heart infusion and casein peptone that supply nitrogenous compounds and amino acids required for the growth of streptococci. Dextrose is an energy source. Bacterial growth is enhanced by yeast extract which supplies essential B vitamins.

  • Ready to use – Continence of prepared media.
  • Selective media – Sodium chloride (6.5%) interferes with the membrane permeability and osmotic equilibrium and inhibits the growth of salt-intolerant bacteria

Easy to interpret -Enterococci grow in the presence of 6.5% NaCl and utilize dextrose in the medium. The metabolism of dextrose produces acid, which causes the indicator, brom cresol purple, to change from purple to yellow.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Heart Infusion Broth w/6.5% NaCl
Format 15mm x 103mm Tube
Product Type Prepared Microbiology Tubes
Quantity 20 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.