Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Horse Defibrinated Blood

Catalog No. R54092 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R54092 100 mL
R54084 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R54092 Supplier Thermo Scientific™ Supplier No. R54092
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Remel Defibrinated Horse Blood is packaged in various sizes.

Thermo Scientific™ Remel Defibrinated Horse Blood is packaged in various sizes.

TRUSTED_SUSTAINABILITY

Specifications

Species Horse
Form Liquid
Product Type Horse Whole Blood
Quantity 100 mL
Processing Defibrinated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.