Learn More
Thermo Scientific™ Remel™ ImmunoWELL™ Mycoplasma IgM Test
Catalog No. R243130 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
96 Wells
Catalog No. Quantity
R243130 96 Wells
1 options
This item is not returnable. View return policy
Catalog No. R243130 Supplier Thermo Scientific™ Supplier No. R243130
Add to Cart
Add to Cart
This item is not returnable. View return policy
An EIA microwell test used for qualitative detection of IgM antibodies to M. pneumoniae in serum.
Remel ImmunoWELL Mycoplasma IgM is an EIA microwell test used for qualitative detection of IgM antibodies to M. pneumoniae in serum.
GenBio
TRUSTED_SUSTAINABILITY
Specifications
Description ImmunoWELL™ Mycoplasma IgM
Detectable Analytes M. pneumoniae IgM
Packaging Type 1 Kit
Quantity 96 Wells