Description
Thermo Scientific™ Remel Inhibitory Mold Agar Bactiflask (with chloramphenicol) is used for primary selective isolation of Histoplasma capsulatum, dermatophytes and other pathogenic fungi.
Specifications
Specifications
| Description | Inhibitory Mold Agar Bactiflask |
| Format | 1 oz. TCF |
| Product Type | Agar |
| Quantity | 10/Pk. |