Learn More
Thermo Scientific™ JEMBEC System with Thayer Martin Agar Improved (M-L) w/anisomycin, colistin, vancomycin, trimethoprim
Catalog No. R10320 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
JEMBEC Plate
Catalog No. Format
R10320 JEMBEC Plate
1 options
This item is not returnable. View return policy
Catalog No. R10320 Supplier Thermo Scientific™ Supplier No. R10320
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Isolate Neisseria gonorrhoeae and Neisseria meningitidis using this medium.
Selectively isolate Neisseria gonorrhoeae and Neisseria meningitidis with Thermo Scientific™ Remel JEMBEC System with Thayer Martin Agar Improved (M-L) w/anisomycin, colistin, vancomycin and trimethoprim. Johnston introduced a medium to grow N. gonorrhoeae in 24 hours1 and this was the advent of GC Agar Base. Various studies have demonstrated improved recovery for pathogenic Neisseria spp. using selective media2-4. Martin and Lewis recommended the use of anisomycin, which is a more stable antifungal agent, to replace nystatin and increase the shelf life of the media5.
Thayer and Martin formulated a selective medium to isolate N. gonorrhoeae and N. meningitidis containing antibiotics capable of suppressing the commensal microbial flora present in the anal canal, vagina, pharynx, and other sites 6. The antibiotics used initially, ristocetin and polymyxin B, were replaced with vancomycin, colistimethate, and nystatin7. The medium contains an increased concentration of dextrose which allows better growth of gonococci.
  • Ready-to-use – Convenience of prepared medium.
  • Selective media –
    • Anisomycin suppresses Candida albicans.
    • Colistin inhibits most gram-negative microorganisms, including Pseudomonas spp.
    • Vancomycin inhibits gram-positive cocci.
    • Trimethoprim lactate suppresses swarming of Proteus spp.
Thayer and Martin Agar (Improved) contains hemoglobin, which provides the X factor (hemin), and GCHI Enrichment that provides the V factor, vitamins, amino acids, coenzymes, and dextrose. The tablet provided in the JEMBEC™ and Pill Pocket systems comes in contact with the water vapor present in the medium and liberates carbon dioxide into the enclosed pack.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description JEMBEC System w/Thayer Martin Agar Improved (Martin-Lewis) w/anisomycin, colistin, vancomycin, trimethoprim
Format JEMBEC Plate
Product Type Agar Plate
Quantity 10/Pk.