Learn More
Thermo Scientific™ Kligler Iron Agar (Slant)

Description
Differentiate enteric gram-negative bacilli on the basis of dextrose and lactose fermentation and hydrogen sulfide (H2S) production using KIA.
Differentiate enteric gram-negative bacilli based on dextrose and lactose fermentation and hydrogen sulfide (H2S) production using Thermo Scientific™ Remel™ Kligler Iron Agar (KIA). In 1918, Klingler described a medium for detection of H2S and differentiation of Salmonella spp.1 Edwards and Ewing recommended KIA for determination of H2S production by enteric gram-negative bacilli2. KIA is also recommended by Gilardi for detection of H2S produced by some strains of Pseudomonas3.
Bailey and Lacey modified the medium by substituting Phenol red indicator for Adrade indicator4. Fermentation reactions are read on the slant and in the butt. Phenol red is an indicator of carbohydrate fermentation whereas, Ferric ammonium citrate is an indicator of H2S production.
- Ready to use – Solid media
- Recommended for determination of H2S production by enteric gram-negative bacilli2 and for detection of H2S produced by some strains of Pseudomonas3
- Easy to interpret
- Carbohydrate Fermentation
- Positive test for Slant/Butt Reaction – Yellow (acid)
- Negative test for Slant/Butt Reaction – Red (alkaline)
- KIA Color Reactions
- Red slat/yellow butt – dextrose (+), lactose (-)
- Yellow slant/yellow butt – dextrose (+), lactose (+)
- Red slant/red butt – dextrose (-), lactose (-)
- Hydrogen Sulphide Production
- Positive test – Black color throughout medium, a black ring at the juncture of the slant and butt, or a black precipitate in the butt
- Negative test – No black color development
- Gas Production
- Positive test – Bubbles in the medium, cracking and displacement of medium, from side and bottom of tube
- Negative test – No bubbles and no separation or displacement of the medium
- Carbohydrate Fermentation
This medium contains casein and meat peptones that supply nitrogenous compounds, amino acids and vitamins required for bacterial growth. Sodium chloride provides essential electrolytes and maintains osmotic equilibrium. The carbohydrate sources present in the medium are lactose and dextrose. Phenol red is an indicator of carbohydrate fermentation whereas ferric ammonium citrate is an indicator of H2S production.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: BAM, COMPF. |
| Description | Kligler Iron Agar (KIA) |
| Format | 15mm x 103mm Tube - Slant |
| Product Type | Agar Slants |
| Quantity | 20/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.