Description
Microscopically examine fungi in wet mount preparations using Lactophenol Aniline Blue.
Microscopically examine fungi in wet mount preparations using Thermo Scientific™ Remel™ Lactophenol Aniline Blue. Linder recommended lactophenol as a mounting fluid for examination and preservation of fungi1. He reported that lactophenol kills organisms without causing cytoplasmic plasmolysis. Lactophenol Aniline Blue is recommended by Koneman, Roberts, and Wright for direct mounting and staining of fungi.2
Lactophenol Aniline Blue is a stain recommended for use in wet mount preparations for direct mounting and staining of fungi.
- Ready to use: Liquid reagent
- Easy identification: Yeast cells, mycelia and fruiting structures stain blue and are visible under a microscope
This reagent contains phenol which precipitates cytoplasmic proteins and inactivates enzyme systems within fungal cell. Glycerol prevents drying, allowing for examination of the stain 18-24 hours after preparation. Lactic acid is a clearing agent. Aniline blue is a dye which stains the hyaline fungal structures making them more distinct.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 25 mL/Bottle |
| Volume (Metric) | 25 mL/Bottle |
| Packaging | Each |