Learn More
Description
Rapidly detect leucine aminopeptidase activity in streptococci and related genera using LAP Disk with Reagent.
Rapidly detect leucine aminopeptidase activity in streptococci and related genera using Thermo Scientific™ Remel™ LAP Disk with Reagent. In 1984, Colman and Ball reported that, unlike streptococci, Aerococcus species do not produce leucine aminopeptidase (LAP)1. Aerococcus strains were found to be consistently negative for leucine aminopeptidase, while strains of Enterococcus and Streptococcus gave positive reactions. LAP, in conjunction with PYR and other conventional tests, is an important parameter in the identification of Aerococcus, Leuconostoc, Pediococcus, and related genera2-4.
Leucine aminopeptidases (LAPs) are a diverse set of exopeptidases that catalyze the hydrolysis of leucine residues from the amino termini of proteins or peptides. LAPs are ubiquitous enzymes present among animals, plants, prokaryotes and microbes. The LAP test should be performed on pure or isolated cultures of catalase-negative, Gram-positive cocci which are 18-72 hours old.
- Ready to use: Reagent-impregnated disc.
- Rapid test: Results are obtained within 5 to 6 minutes.
- Easy to identify: A color change from pink to red indicates a positive test whereas no color change or a slightly yellow color indicates a negative test.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Murex Lap Kit/50/Pk. |
| CE Marker | Yes |
| Includes | LAP disk and color developer included |
| Quantity | 50 Tests/Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
