Learn More
Thermo Scientific™ Remel™ LB Agar w/Ampicillin (100μg/mL) (Luria-Bertani)
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Remel LB Agar with Ampicillin (100μg/mL) (Luria-Bertani) is used for the cultivation and maintenance of Escherichia coli for genetic and molecular studies.
Specifications
Specifications
| Description | LB (Luria-Bertani) Agar w/Ampicillin 100micrograms/mL |
| Format | 15mm x 150mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |