Description
Thermo Scientific™ Remel LB Agar with Tetracycline, 25μg/mL (Luria-Bertani) is for cultivation and maintenance of Escherichia coli for genetic and molecular studies.
Specifications
Specifications
| Description | LB Agar w/Tetracycline, 25µg |
| Format | Monoplate Deep Pour |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |