Learn More
Thermo Scientific™ Light Green Counterstain
Catalog No. R40123 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
R40123 Each
1 options
This item is not returnable. View return policy
Catalog No. R40123 Supplier Thermo Scientific™ Supplier No. R40123
This item is not returnable. View return policy
Use Light Green as a counterstain in the modified acid-fast procedure to detect Cryptosporidium in clinical specimens.
Use Thermo Scientific™ Remel™ Light Green Counterstain to detect Cryptosporidium in clinical specimens.
Cryptosporidium is an intestinal protozoan parasite that causes diarrhea in animals as well as humans. This strain causes infection in immunocompromised hosts, especially those with AIDS (Acquired Immune Deficiency Syndrome)1. A modified acid-fast stain procedure was recommended by Ma and Soave because of its simplicity and specificity to detect Cryptosporidium. Recent modification of the Kinyoun carbolfuchsin stain is recommended by Garcia for detection of Cryptosporidium oocysts in clinical specimens2.
Cryptosporidium oocysts are spherical 4-6 μm in diameter, and stain pink to red to purple with Kinyoun carbolfuchsin stain. Most will contain several dark staining granules. When light green is used as a counterstain, yeast cells, which are morphologically similar to Cryptosporidium oocysts, stain green3.
Cryptosporidium is more difficult to detect in formed stool specimens. Stain time is increased that allows the oocysts to stand out from the background material. The hot acid-fast stain maximizes detection and indentification of Cryptosporidium in formed stools3.
  • Easy-to-use - Examine smear in the slides mounted with mounting medium and a coverslip by bright field microscopy (40 X objective) and oil immersion is used (100 X objective) to observe the internal morphology.
  • Easy-to-interpret - In case of positive test oocysts stain pink to red to purple, are 4-6 μm in diameter and round or oval in shape. One to four sporozoites maybe visible within the oocyst. In case of negative test the yeast cells and background material stain green in color.
Light Green Counterstain contains Light Green (CAS 5141-20-8), Glacial Acetic Acid (CAS 64-19-7) and Demineralized Water (CAS 7732-18-5).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Quantity Each
Volume (Metric) 250 mL/Bottle
Product Type Counterstain