Learn More
Thermo Scientific™ L-J Gruft
GSA_VA
Catalog No. R08520 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
1 oz. Bottle
20mm x 113mm Tube - Slant
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R08520 100/Pk. 20mm x 113mm Tube - Slant
R08522 20/Pk. 20mm x 113mm Tube - Slant
R10103 10/Pk. 1 oz. Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R08520 Supplier Thermo Scientific™ Supplier No. R08520
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Selectively isolate and cultivate Mycobacterium spp. using Lowenstein-Jensen (L-J) Medium, L-J Gruft, and L-J Mycobactocel.
Selectively isolate and cultivate Mycobacterium spp. using Thermo Scientific™ Remel™ Lowenstein-Jensen (L-J) Medium, L-J Gruft, and L-J Mycobactocel. Medium consisting of congo red and malachite green dyes was formulated by Lowenstein to cultivate mycobacteria1. This medium was modified by Jensen by replacing congo red with citrate and phosphate contents and increasing the malachite green concentration2. L-J Medium used for recovery of pathogenic Mycobacterium spp. was found to be superior to other media tested, including ATS (American Trudeau Society) Medium3,4.
Selectively isolate and cultivate Mycobacterium spp. using Lowenstein-Jensen (L-J) Medium, L-J Gruft, and L-J Mycobactocel.
  • Selective – Antibiotics in the medium make the medium selective for Mycobacterium while inhibiting contaminating bacteria.
  • Ready to use – Convenience of prepared media.
L-J Medium is a glycerated egg-potato medium. The glycerol and egg mixture provide fatty acids and protein required for the metabolism of mycobacteria. L-J Medium was modified by Gruft by adding ribonucleic acid as a growth stimulant and penicillin and nalidixic acid as selective inhibitors of contaminating bacteria5. Ribonucleic acid increases the percentage of positive cultures over the standard L-J Medium. The selectivity of L-J Medium was increased by incorporating cycloheximide, lincomycin, and nalidixic acid and was formulated by Petran and Vera6.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Lowenstein-Jensen Gruft. (LJ) w/penicillin, nalidixic acid
Format 20mm x 113mm Tube - Slant
Product Type Prepared Microbiology Tubes
Quantity 100/Pk.