Learn More
Thermo Scientific™ Remel™ Lowenstein-Jensen Medium w/2.5% Ferric Ammonium Citrate
Shop All Thermo Scientific Products
Description
Thermo Scientific™ Remel Lowenstein-Jensen Medium with 2.5% Ferric Ammonium Citrate is for the iron uptake reaction by Mycobacterium spp.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Lowenstein-Jensen Medium w/5% Ferric Ammonium Citrate |
| Format | 20mm x 113mm Tube - Slant |
| Product Type | Microbiology Media |
| Quantity | 20/Pk. |