Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lysozyme Control Broth
GSA_VA

Catalog No. R060972 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R060972 20 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R060972 Supplier Thermo Scientific™ Supplier No. R060972
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differentiate aerobic actinomycetes using Lysozyme Control Broth.

Differentiate aerobic actinomycetes like Nocardia and Streptomycesusing Thermo Scientific™ Remel™ Lysozyme Control Broth. A taxonomic scheme for identification of aerobic actinomycetes was developed based on biochemical characteristics which included testing for the growth in the presence of lysozyme1. The enzyme lysozyme breaks down the cell walls of certain aerobic actinomycetes such as Streptomyces spp. and are referred sensitive and will not grow in lysozyme broth. Lysozyme resistance is more reliable to differentiate Nocardia and Streptomyces2.

The aerobic actinomycetes are a heterogenous group of genera and include species of Nocardia and Streptomyces3. As a group, the organisms are gram-positive, catalase-positive that may be presumptively identified by staining properties, microscopic morphology, and substrate degradation4. Most Nocardia spp. are resistant and grow in the presence of lysozyme.

This medium contains beef extract, peptone, and glycerol that supply carbohydrates, vitamins, nitrogen compounds, salts, and amino acids necessary for the growth of aerobic actinomycetes.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Lysozyme Control Broth
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.