Learn More
Thermo Scientific™ m ENDO Agar LES
Catalog No. R01426 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
Catalog No. Format
R01426 12mm x 85mm Monoplate
1 options
This item is not returnable. View return policy
Catalog No. R01426 Supplier Thermo Scientific™ Supplier No. R01426
Add to Cart
Add to Cart
This item is not returnable. View return policy
Recommended for cultivation and enumeration of coliform bacteria from water by membrane filtration method.
Cultivate and enumerate coliform bacteria from water by membrane filtration method using Thermo Scientific™ Remel™ m ENDO Agar LES (Dehydrated). The m ENDO Agar LES medium is prepared according to the formulation of McCarthy, Delaney, and Grasso1 and used for the enumeration of coliform organisms in water2. It is recommended by the American Public Health Association (APHA) in Standard Methods for the Examination of Water and Wastewater and Compendium of Methods for the Microbiological Examination of Foods for the enumeration of coliforms in water, wastewater, and foods2,3.
Coliforms are indicator organisms used for assessing the effectiveness of water treatment and disinfection, and for monitoring the sanitary integrity of water and wastewater. They are aerobic or facultative anaerobic, gram-negative, non-spore forming b
TRUSTED_SUSTAINABILITY
Specifications
Description m-ENDO Agar LES
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.