Learn More
Thermo Scientific™ MacConkey Agar with Sorbitol
Catalog No. R09548
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R09548 20/Pk.
R01556 10/Pk.
R01561 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R09548 Supplier Thermo Scientific™ Supplier No. R09548
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selectively detect sorbitol negative Escherichia coli from clinical specimens with this selective and specific medium.
Selectively detect sorbitol negative Escherichia coli from clinical specimens with Thermo Scientific™ Remel™ MacConkey Agar with Sorbitol prepared media plates. The medium is recommended for the isolation of verotoxin-producing E.coli O157: H7. Unlike most serotypes of E.coli, verotoxin-producing E.coli O157: H7 does not ferment sorbitol and, therefore, produces colorless colonies. In contrast, most E.coli strains ferment sorbitol and form pink colonies.
Escherichia coli O157 is a significant human pathogen and is recognized as a cause of hemorrhagic colitis, an illness characterized by bloody diarrhea, severe abdominal pain, and hemolytic uremic syndrome.
Use Sorbitol MacConkey Agar for selective detection of Escherichia coli O157.
  • Sensitive and specific for E. coli O157 based on sorbitol fermentation.
  • Recommended as a simple, inexpensive, rapid and reliable means of screening for E. coli O157.
  • Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: BAM, COMPF.
Description MacConkey Agar w/Sorbitol
Format 20mm x 150mm Pour Tube
Product Type Agar Plate
Quantity 20/Pk.