Learn More
Thermo Scientific™ MacConkey Agar with Sorbitol

Description
Selectively detect sorbitol negative Escherichia coli from clinical specimens with Thermo Scientific™ Remel™ MacConkey Agar with Sorbitol prepared media plates. The medium is recommended for the isolation of verotoxin-producing E.coli O157: H7. Unlike most serotypes of E.coli, verotoxin-producing E.coli O157: H7 does not ferment sorbitol and, therefore, produces colorless colonies. In contrast, most E.coli strains ferment sorbitol and form pink colonies.
Escherichia coli O157 is a significant human pathogen and is recognized as a cause of hemorrhagic colitis, an illness characterized by bloody diarrhea, severe abdominal pain, and hemolytic uremic syndrome.
Use Sorbitol MacConkey Agar for selective detection of Escherichia coli O157.
- Sensitive and specific for E. coli O157 based on sorbitol fermentation.
- Recommended as a simple, inexpensive, rapid and reliable means of screening for E. coli O157.
- Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: BAM, COMPF. |
| Description | MacConkey Agar w/Sorbitol |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.