Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Malonate Broth,Ewing

Catalog No. R061326 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R061326 20 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R061326 Supplier Thermo Scientific™ Supplier No. R061326
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differentiate members of Enterobacteriaceae based on sodium malonate utilization using Malonate Broth (Ewing Modification).

Differentiate members of Enterobacteriaceae based on sodium malonate utilization using Thermo Scientific™ Remel™ Malonate Broth (Ewing Modification). Malonate Broth is recommended to differentiate certain subgroups of Salmonella from other Enterobacteriaceae1. Leifson developed a liquid medium to differentiate Escherichia coli from Enterobacter spp2. Ewing et al. modified the Leifson's formulation by adding a trace amount of dextrose and yeast extract to stimulate growth of certain organsims that fail to grow and obseved their reaction on malonate3,4.

Differentiate members of Enterobacteriaceae based on sodium malonate utilization using Malonate Broth (Ewing Modification).

  • Recommended to differentiate certain subgroups of Salmonella from other Enterobacteriaceae1
  • Blue color development due to utilization of malonate is easy to interpret
  • Ready to use – Convenience of prepared media

An organism that utilizes sodium malonate as a carbon source and ammonia sulfate as a nitrogen source produces an alkaline reaction and organisms which cannot utilize malonate will ferment dextrose, causes the pH indicator to change from green to yellow thus producing an acid reaction.

TRUSTED_SUSTAINABILITY

Specifications

Description Malonate Broth
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.